missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Clinofibrate, TCI America™

Manufacturer:  TCI America C31001G

 View more versions of this product

Catalog No. C31001G



Specifications

Specifications

Clinofibrate
145°C
30299-08-2
MFCD00865769
BMOVQUBVGICXQN-UHFFFAOYSA-N
2-[4-[1-[4-(2-carboxybutan-2-yloxy)phenyl]cyclohexyl]phenoxy]-2-methylbutanoic acid
2787
Crystal-Powder at 20°C
White-Yellow
1g
C28H36O6
2,2′C-(4,4′C-Cyclohexylidenediphenoxy)-2,2′C-dimethyldibutanoic Acid
CCC(C)(C(=O)O)OC1=CC=C(C=C1)C2(CCCCC2)C3=CC=C(C=C3)OC(C)(CC)C(=O)O
468.59
468.59
≥98.0% (HPLC)

Chemical Identifiers

30299-08-2
468.59
BMOVQUBVGICXQN-UHFFFAOYSA-N
2787
CCC(C)(C(=O)O)OC1=CC=C(C=C1)C2(CCCCC2)C3=CC=C(C=C3)OC(C)(CC)C(=O)O
C28H36O6
MFCD00865769
2,2′C-(4,4′C-Cyclohexylidenediphenoxy)-2,2′C-dimethyldibutanoic Acid
2-[4-[1-[4-(2-carboxybutan-2-yloxy)phenyl]cyclohexyl]phenoxy]-2-methylbutanoic acid
Safety and Handling

Safety and Handling

RTECSNumber : ES8900000

TSCA : No

Documents