missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Clindamycin phosphate

Antibiotic U-28508E, effective against Gram-positive bacteria

$363.95 - $363.95

Chemical Identifiers

CAS 24729-96-2
Molecular Formula C18H34ClN2O8PS
Molecular Weight (g/mol) 504.96
MDL Number MFCD07793328
InChI Key UFUVLHLTWXBHGZ-AVENPWRCSA-N
PubChem CID 71314904
IUPAC Name [(2R,3R,4S,5R,6R)-6-[(1S,2R)-2-chloro-1-[[(2S,4R)-1-methyl-4-propylpyrrolidine-2-carbonyl]amino]propyl]-4,5-dihydroxy-2-methylsulfanyloxan-3-yl] dihydrogen phosphate
SMILES CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)OP(=O)(O)O)O)O)C(C)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63387MC View Documents Alfa Aesar™
J63387MC
100mg Each for $363.95
 
Specifications

Chemical Identifiers

24729-96-2
504.96
UFUVLHLTWXBHGZ-AVENPWRCSA-N
[(2R,3R,4S,5R,6R)-6-[(1S,2R)-2-chloro-1-[[(2S,4R)-1-methyl-4-propylpyrrolidine-2-carbonyl]amino]propyl]-4,5-dihydroxy-2-methylsulfanyloxan-3-yl] dihydrogen phosphate
C18H34ClN2O8PS
MFCD07793328
71314904
CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)OP(=O)(O)O)O)O)C(C)Cl

Specifications

24729-96-2
MFCD07793328
Freely soluble in water
CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)OP(=O)(O)O)O)O)C(C)Cl
504.96
504.96
White
100mg
C18H34ClN2O8PS
14,2356
UFUVLHLTWXBHGZ-AVENPWRCSA-N
[(2R,3R,4S,5R,6R)-6-[(1S,2R)-2-chloro-1-[[(2S,4R)-1-methyl-4-propylpyrrolidine-2-carbonyl]amino]propyl]-4,5-dihydroxy-2-methylsulfanyloxan-3-yl] dihydrogen phosphate
71314904
Solid
114°C
Clindamycin phosphate
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : GF2625000