Please login to your online account to display your discounted pricing

Clindamycin Hydrochloride, Tocris™

Antibiotic; inhibits protein synthesis

$95.70 - $401.94

Chemical Identifiers

Chemical Name or Material Clindamycin Hydrochloride
Assay Percent Range >98%
Molecular Formula C18H33ClN2O5S·HCl
Formula Weight 461.44
Synonym Methyl-7-chloro-6,7,8-trideoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-L-threo-α-D-galacto-octopyranoside hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

482210

 
r & d systems
RDS482210
10mg Each for $95.70

482250

 
r & d systems
RDS482250
50mg Each for $401.94
Description & Specifications

Specifications

Formula Weight 461.44
Quantity 10mg
Molecular Formula C18H33ClN2O5S·HCl
CAS 21462-39-5
Synonym Methyl-7-chloro-6,7,8-trideoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1-thio-L-threo-α-D-galacto-octopyranoside hydrochloride
Recommended Storage Store at -20°C
Chemical Name or Material Clindamycin Hydrochloride
Assay Percent Range >98%
Safety and Handling

Store at -20°C