missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Clemizole Hydrochloride, TCI America™

Manufacturer:  TCI America C3468100MG

 View more versions of this product

Catalog No. C3468100MG



Specifications

Specifications

Clemizole Hydrochloride
241°C
1163-36-6
MFCD00051435
DNFMJYXRIMLMBZ-UHFFFAOYSA-N
1-[(4-chlorophenyl)methyl]-2-(pyrrolidin-1-ylmethyl)benzimidazole;hydrochloride
5309446
Crystal-Powder at 20°C
White
100mg
C19H21Cl2N3
1-(4-Chlorobenzyl)-2-(pyrrolidin-1-ylmethyl)benzimidazole Hydrochloride
C1CCN(C1)CC2=NC3=CC=CC=C3N2CC4=CC=C(C=C4)Cl.Cl
362.298
362.30
≥98.0% (HPLC)

Chemical Identifiers

1163-36-6
362.298
DNFMJYXRIMLMBZ-UHFFFAOYSA-N
5309446
C1CCN(C1)CC2=NC3=CC=CC=C3N2CC4=CC=C(C=C4)Cl.Cl
C19H21Cl2N3
MFCD00051435
1-(4-Chlorobenzyl)-2-(pyrrolidin-1-ylmethyl)benzimidazole Hydrochloride
1-[(4-chlorophenyl)methyl]-2-(pyrrolidin-1-ylmethyl)benzimidazole;hydrochloride
Safety and Handling

Safety and Handling

RTECSNumber : DD6730000

TSCA : No

Recommended Storage : Refrigerator

Documents