Please login to your online account to display your discounted pricing

Qorpak™ Clear Standard Wide Mouth Bottles with Phenolic Pulp/Tin Foil Lined Cap

Provides maximum visibility and integrity of the sample contents. Qorpak™ Clear Standard Wide Mouth Bottles with Phenolic Pulp/Tin Foil Lined Cap are ideal for general purpose.

$100.77 - $100.77

Specifications

Closure Liner Pulp/Tin Foil
Closure Material Phenolic
Closure Screw
Closure Color Black
Color Clear
View More Specs
Products
Catalog Number Mfr. No. Capacity Height (English) Packaging Type Quantity Price Quantity    

02-993-054

 
qorpak™
GLC01753
4 oz., 120mL 3.39 in. Conventional Pack 24/Cs. Case for $100.77
Description & Specifications

Specifications

Closure Liner Pulp/Tin Foil
Closure Material Phenolic
Closure Screw
Closure Color Black
Color Clear
Material Glass

  • Features wide mouth opening
  • Closures offer good chemical resistance and are typically used with solids and powders
  • Resistant to hydro-carbons, alcohols, ketones and oils
  • Not for acids and alkalis
  • Applications include pharmaceutical, chemical, environmental and food packaging and storing