Please login to your online account to display your discounted pricing
Fisherbrand™ Safety Coated Clear Boston Round Bottles with Black Phenolic PolyCone Cap
General use bottle ideal when content integrity is important. Fisherbrand™ Safety Coated Clear Boston Round Bottles with Black Phenolic PolyCone Cap help to insure product integrity for long term storage.
$132.33 - $132.33
Specifications
Closure Material Phenolic
Closure Liner Polyethylene
Shape Boston Round
Mouth Narrow
View More Specs
Products
Catalog Number Mfr. No. Neck Finish Capacity (Metric) Price Quantity    
02-911-971
fisherbrand™
263880
28-400 16 oz., 480mL Case of 12 Each for $132.33
Description & Specifications
Specifications
Closure Material Phenolic
Closure Liner Polyethylene
Shape Boston Round
Mouth Narrow
  • Glass packers coated in plastisol, which is a solution of PVC resin in a liquid plasticizer that cools to form a perfectly-fitting plastic coating
  • Plastic coating contains the substance in the bottle long enough to allow for proper disposal if the glass breaks
  • Harder to break and easier to grip thus reducing breakage