Clear Polystyrene 96-Well Microplates, 330µL, With Lid
Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ Clear Polystyrene 96-Well Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 96-well microplates for laboratory miniaturization and automation.
$865.62 - $1,375.89
Products 2
Catalog Number Mfr. No. Packaging Price Quantity  
Catalog Number Mfr. No. Packaging Price Quantity  
07-200-720A View Documents Corning™
C3358
Bags of 20 Case of 100 Each for $1,375.89
07-200-95 View Documents Corning™
C3799
Individually Wrapped Case of 50 Each for $865.62
Description
Description
  • Medical-grade polystyrene
  • Treated surface options are not coated, require no special handling and are stable at room temperature
  • Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
  • Ideal for serial dilutions, hemagglutination or precipitation assays, and general storage applications
  • Plate dimensions meet proposed industry standards