missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Qorpak™ Clear Molded Glass Type I Borosilicate Serum Vials
Description
- 100% inspected in clean room environment
- Vials are washed with water for injection and dried in an ISO8 environment
- Vials are depyrogenated in a tunnel according to European and US pharmaccopeias
- Vials are placed in trays for packaging in ISO5 area with ISO7 surrounding
- Sealed with a Tyvek™ lid in ISO5 environment, packed in steribags and placed into abox allowing for sterilization in ISO8 surrounding
- Sterilization is made through ethylene oxide and a process validated according to the ISO11135 normal
Specifications
Specifications
| Color | Clear |
| Diameter (Metric) Outer | 32 mm |
| Material | Borosilicate Glass |
| Pre-slit | No |
| Sterility | Sterile |
| Disposable | Single-use |
| Volume (Metric) | 20 mL |
| For Use With (Application) | Lyophilization |
| Graduated | No |
| No. per Case | 480/Cs. |
| Show More |