Fisherbrand™ Clear Jug with Black Phenolic PolyCone Cap
Ideal for storage for solids, liquids, powders and specimens.
Manufacturer:  Fisherbrand™ 278401
 View more versions of this product
Catalog No. 02-912-272A
Description
Description
  • Wide-mouth design for efficient addition and removal of contents
  • Clear glass allows for easy viewing of contents
  • Graduated in both milliliters and ounces
Specifications
Specifications
3840mL
Phenolic
Clear
Glass
Black Ribbed
38-400
128 oz.
Polycone
Polycone Cap
Jug
Yes