Fisherbrand™ Clear Graduated Glass Bottles with Polypropylene Pulp/Vinyl Lined Cap
Multipurpose containers for use with biological and pathological specimens. Fisherbrand™ Clear Graduated Glass Bottles with Polypropylene Pulp/Vinyl Lined Caps can also be used for storing industrial laboratory chemicals and reagents.
$37.37 - $373.74
Specifications
Liner Material Pulp/Vinyl
Closure Material Polypropylene
Closure Type Screw
Closure Color White
Color Clear
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Neck Finish Quantity Capacity (English) Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Neck Finish Quantity Capacity (English) Capacity (Metric) Price Quantity  
02-911-453A Fisherbrand™
ALP263470
33-400 48/Cs. 1 oz. 30mL Case of 48 Each for $44.31
02-911-454A Fisherbrand™
ALP263478
38-400 48/Cs. 2 oz. 60mL Case of 48 Each for $62.53
02-911-456A Fisherbrand™
ALP263489
58-400 24/Cs. 8 oz. 240mL Case of 24 Each for $78.13
02-911-455A Fisherbrand™
ALP263484
48-400 24/Cs. 4 oz. 120mL Case of 24 Each for $37.37
02-911-457A Fisherbrand™
ALP263461
70-400 24/Cs. 16 oz. 480mL Case of 24 Each for $83.30
02-911-465A Fisherbrand™
ALP263469
33-400 432/Cs. 1 oz. 30mL Case of 432 Each for $373.74
02-911-466A Fisherbrand™
ALP263477
38-400 288/Cs. 2 oz. 60mL Case of 288 Each for $295.00
02-911-467A Fisherbrand™
ALP263483
48-400 144/Cs. 4 oz. 120mL Case of 144 Each for $186.57
02-911-469A Fisherbrand™
ALP263462
70-400 48/Cs. 16 oz. 480mL Case of 48 Each for $150.90
02-911-468A Fisherbrand™
ALP263490
58-400 96/Cs. 8 oz. 240mL Case of 96 Each for $219.86
Specifications
Description
  • Graduated in both milliliters and ounces
  • Clear (flint) glass bottles are designed with a slight shoulder for easy pouring and handling
  • Wide mouth facilitates easy mixing, handling, and transfer of contents
White Polypropylene Pulp/Vinyl Lined Caps
  • Polypropylene Caps are known for good impact strength, durability, cost effectiveness, thermal resistance, and pliability
  • Caps are autoclavable
  • Liner is made of a vinyl coating applied to high density polyethylene coated paper and then it is laminated to pulpboard
Description & Specifications
Specifications
Pulp/Vinyl
Screw
Clear
Wide
Polypropylene
White
Glass
Round