missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Clear Graduated Glass Bottles with Polypropylene Pulp/Vinyl Lined Cap

Multipurpose containers for use with biological and pathological specimens. Fisherbrand™ Clear Graduated Glass Bottles with Polypropylene Pulp/Vinyl Lined Caps can also be used for storing industrial laboratory chemicals and reagents.

Manufacturer:  Fisherbrand™ ALP263462

 View more versions of this product

Catalog No. 02-911-469A


 

Includes: Polypropylene Pulp/Vinyl lined cap

Specifications

Description

AC Round, Graduated; Fisherbrand; Clear; with white Polypropylene Pulp/Vinyl lined cap attached; 16 oz., 480mL; 70-400; 48/Cs.

  • Graduated in both milliliters and ounces
  • Clear (flint) glass bottles are designed with a slight shoulder for easy pouring and handling
  • Wide mouth facilitates easy mixing, handling, and transfer of contents

White Polypropylene Pulp/Vinyl Lined Caps
  • Polypropylene Caps are known for good impact strength, durability, cost effectiveness, thermal resistance, and pliability
  • Caps are autoclavable
  • Liner is made of a vinyl coating applied to high density polyethylene coated paper and then it is laminated to pulpboard
Description & Specifications

Specifications

480mL
Polypropylene
Glass
Bottle, Graduated
Pulp/Vinyl
70-400
Screw
16 oz.
Clear
48/Cs.
White
Wide
Round