Please login to your online account to display your discounted pricing

Fisherbrand™ Clear Graduated Glass Bottles with Polypropylene Pulp/Vinyl Lined Cap


Multipurpose containers for use with biological and pathological specimens. Fisherbrand™ Clear Graduated Glass Bottles with Polypropylene Pulp/Vinyl Lined Caps can also be used for storing industrial laboratory chemicals and reagents.

Manufacturer: fisherbrand™  ALP263489

 View more versions of this product

Polypropylene Pulp/Vinyl lined cap

Catalog No. 02-911-456A

  • / Case of 24 Each
 

Description & Specifications

Specifications

Closure Color White
Type Graduated Bottle
Liner Material Pulp/Vinyl
Closure Type Screw
Mouth Wide
Neck Finish 58-400
Shape Round
Capacity (Metric) 240mL
Closure Material Polypropylene
Material Glass
Quantity 24/Cs.
Color Clear
Capacity (English) 8 oz.

  • Graduated in both milliliters and ounces
  • Clear (flint) glass bottles are designed with a slight shoulder for easy pouring and handling
  • Wide mouth facilitates easy mixing, handling, and transfer of contents

White Polypropylene Pulp/Vinyl Lined Caps
  • Polypropylene Caps are known for good impact strength, durability, cost effectiveness, thermal resistance, and pliability
  • Caps are autoclavable
  • Liner is made of a vinyl coating applied to high density polyethylene coated paper and then it is laminated to pulpboard