Please login to your online account to display your discounted pricing
Fisherbrand™ Clear Graduated Glass Bottles with Polypropylene Pulp/Vinyl Lined Cap
Multipurpose containers for use with biological and pathological specimens. Fisherbrand™ Clear Graduated Glass Bottles with Polypropylene Pulp/Vinyl Lined Caps can also be used for storing industrial laboratory chemicals and reagents.
Manufacturer: fisherbrand™  ALP263484
 View more versions of this product
Polypropylene Pulp/Vinyl lined cap
Catalog No. 02-911-455A
  • / Case of 24 Each
Description & Specifications
Specifications
Capacity (Metric) 120mL
Capacity (English) 4 oz.
Product Type Graduated Bottle
Closure Color White
Closure Material Polypropylene
Closure Type Screw
Color Clear
Material Glass
Liner Material Pulp/Vinyl
Mouth Wide
Neck Finish 48-400
Quantity 24/Cs.
Shape Round
  • Graduated in both milliliters and ounces
  • Clear (flint) glass bottles are designed with a slight shoulder for easy pouring and handling
  • Wide mouth facilitates easy mixing, handling, and transfer of contents
White Polypropylene Pulp/Vinyl Lined Caps
  • Polypropylene Caps are known for good impact strength, durability, cost effectiveness, thermal resistance, and pliability
  • Caps are autoclavable
  • Liner is made of a vinyl coating applied to high density polyethylene coated paper and then it is laminated to pulpboard