missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Graduated Glass Bottle Beakers with White Polypropylene Pulp/Vinyl Cap

Multipurpose containers for use with biological and pathological specimens. Fisherbrand™ Clear Graduated Glass Bottles with Polypropylene Pulp/Vinyl Lined Caps can also be used for storing industrial laboratory chemicals and reagents.

Manufacturer:  Fisherbrand™ ALP263469

 View more versions of this product

Catalog No. 02-911-465A


 

Includes: Polypropylene Pulp/Vinyl lined cap

Description

Description

Graduated Medium Rounds are excellent multipurpose containers for use with biological and pathological specimens, but can also be used for storing industrial laboratory chemicals and reagents. These clear (flint) glass bottles are designed with a slight shoulder for easy pouring and handling. The wide mouth facilitates easy mixing, handling, and transfer of contents. Graduated in both milliliters and ounces.

  • Graduated in both milliliters and ounces
  • Clear (flint) glass bottles are designed with a slight shoulder for easy pouring and handling
  • Wide mouth facilitates easy mixing, handling, and transfer of contents

White Polypropylene Pulp/Vinyl Lined Caps
  • Polypropylene Caps are known for good impact strength, durability, cost effectiveness, thermal resistance, and pliability
  • Caps are autoclavable
  • Liner is made of a vinyl coating applied to high density polyethylene coated paper and then it is laminated to pulpboard
Specifications

Specifications

30mL
Polypropylene
GlassþGlass
Bottle, GraduatedþGraduated Medium Round Bottle
Pulp/Vinyl
33-400
Screw
1 oz.
ClearþClear
432/Cs.
White
Wide
Round
Documents