Fisherbrand™ Graduated Glass Bottles with Black Phenolic Rubber Cap
Multipurpose containers for use with biological and pathological specimens. Fisherbrand™ Graduated Glass Bottles with Black Phenolic Rubber Cap is also used for storing industrial laboratory chemicals and reagents.
$70.30 - $115.67
Specifications
Liner Material Rubber
Closure Material Phenolic
Closure Type Screw
Closure Color Black
Color Clear
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Neck Finish Quantity Capacity (English) Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Neck Finish Quantity Capacity (English) Capacity (Metric) Price Quantity  
02-911-474A Fisherbrand™
ALP263474
38-400 48/Cs. 2 oz. 60mL Case of 48 Each for $80.40
02-911-473A Fisherbrand™
ALP263466
33-400 48/Cs. 1 oz. 30mL Case of 48 Each for $83.10
02-911-475A Fisherbrand™
ALP263480
48-400 24/Cs. 4 oz. 120mL Case of 24 Each for $70.30
02-911-476A Fisherbrand™
ALP263486
58-400 24/Cs. 8 oz. 240mL Case of 24 Each for $115.67
Specifications
Description
  • Graduated in both milliliters and ounces
  • Feature a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents; clear (flint) glass is appropriate when visibility is important but protection from UV rays is not necessary
  • Environmentally sensitive bottles help eliminate waste and help to insure product integrity for long term storage
Black Phenolic Rubber Lined Caps
  • Autoclavable
  • Liner exhibit good resiliency and resistance to moisture vapor
  • Closures provide consistent adherence to close dimensional tolerance
  • Liner is compliant with FDA 21 CFR 177.2600 Rubber Articles Intended for Repeated Use
Description & Specifications
Specifications
Rubber
Screw
Clear
Wide
Phenolic
Black
Glass
Round