Please login to your online account to display your discounted pricing

Fisherbrand™ Graduated Glass Bottles with Black Phenolic Rubber Cap


Multipurpose containers for use with biological and pathological specimens. Fisherbrand™ Graduated Glass Bottles with Black Phenolic Rubber Cap is also used for storing industrial laboratory chemicals and reagents.

Manufacturer: fisherbrand™  ALP263474

 View more versions of this product

Phenolic Rubber lined cap

Catalog No. 02-911-474A

  • / Case of 48 Each
 

Description & Specifications

Specifications

Closure Color Black
Type Graduated Bottle
Mouth Wide
Closure Type Screw
Neck Finish 38-400
Liner Material Rubber
Shape Round
Capacity 2 oz., 60mL
Graduated Yes
Capacity (Metric) 60mL
Capacity (English) 2 oz.
Quantity 48/Cs.
Material Glass
Color Clear
Closure Material Phenolic

  • Graduated in both milliliters and ounces
  • Feature a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents; clear (flint) glass is appropriate when visibility is important but protection from UV rays is not necessary
  • Environmentally sensitive bottles help eliminate waste and help to insure product integrity for long term storage
Black Phenolic Rubber Lined Caps
  • Autoclavable
  • Liner exhibit good resiliency and resistance to moisture vapor
  • Closures provide consistent adherence to close dimensional tolerance
  • Liner is compliant with FDA 21 CFR 177.2600 Rubber Articles Intended for Repeated Use