Please login to your online account to display your discounted pricing

Fisherbrand™ Graduated Glass Bottles with Black Phenolic Rubber Cap


Multipurpose containers for use with biological and pathological specimens. Fisherbrand™ Graduated Glass Bottles with Black Phenolic Rubber Cap is also used for storing industrial laboratory chemicals and reagents.

Manufacturer: fisherbrand™  ALP263466

 View more versions of this product

Phenolic Rubber lined cap

Catalog No. 02-911-473A

  • / Case of 48 Each
 

Description & Specifications

Specifications

Capacity (Metric) 30mL
Capacity (English) 1 oz.
Product Type Graduated Bottle
Closure Color Black
Closure Material Phenolic
Closure Type Screw
Color Clear
Graduated Yes
Material Glass
Liner Material Rubber
Mouth Wide
Neck Finish 33-400
Quantity 48/Cs.
Shape Round
Capacity 1 oz., 30mL

  • Graduated in both milliliters and ounces
  • Feature a narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents; clear (flint) glass is appropriate when visibility is important but protection from UV rays is not necessary
  • Environmentally sensitive bottles help eliminate waste and help to insure product integrity for long term storage
Black Phenolic Rubber Lined Caps
  • Autoclavable
  • Liner exhibit good resiliency and resistance to moisture vapor
  • Closures provide consistent adherence to close dimensional tolerance
  • Liner is compliant with FDA 21 CFR 177.2600 Rubber Articles Intended for Repeated Use