Please login to your online account to display your discounted pricing

Fisherbrand™ Clear Graduated Glass Bottles with Black Phenolic Pulp/Tin Foil Lined Cap

Clear Glass Boston Round Bottles are excellent general use bottles ideal for when content integrity and sample or product visibility is important.

$51.37 - $107.97

Specifications

Liner Material Pulp/Tin Foil
Closure Material Phenolic
Closure Type Screw
Closure Color Black
Color Clear
View More Specs

 Alert

Not recommended for acids and alkalis.

Products
Catalog Number Mfr. No. Quantity Capacity (English) Capacity (Metric) Neck Finish Price Quantity    

02-911-911A

 
fisherbrand™
ALP263460
24/Cs. 16 oz. 480mL 70-400 Case of 24 Each for $107.97

02-911-923A

 
fisherbrand™
ALP263468
48/Cs. 1 oz. 30mL 33-400 Case of 48 Each for $68.87

02-911-924A

 
fisherbrand™
ALP263476
48/Cs. 2 oz. 60mL 38-400 Case of 48 Each for $86.43

02-911-925A

 
fisherbrand™
ALP263482
24/Cs. 4 oz. 120mL 48-400 Case of 24 Each for $51.37

02-911-926A

 
fisherbrand™
ALP263488
24/Cs. 8 oz. 240mL 58-400 Case of 24 Each for $76.43
Description & Specifications

Specifications

Liner Material Pulp/Tin Foil
Closure Material Phenolic
Closure Type Screw
Closure Color Black
Color Clear
Material Glass
Mouth Wide
Shape Round

Clear Graduated Glass Bottles

  • Graduated in both milliliters and ounces
  • For use with biological and pathological specimens, but can also be used for storing industrial laboratory chemicals and reagents
  • Glass offers minimal chemical interference with contents; clear (flint) glass is appropriate when visibility is important but protection from UV rays is not necessary
  • Designed with a slight shoulder for easy pouring and handling
  • Wide mouth facilitates easy mixing, handling, and transfer of contents
Pulp/Tin-Foil Lined Phenolic Caps
  • Offer good chemical, impact, and temperature resistance and are typically used with solids and powders
  • Plastic screw caps with pulp/tin-foil liners have strong resistance to hydro-carbons, alcohols, ketones and oils, but are not recommended for acids and alkalis