Please login to your online account to display your discounted pricing

Fisherbrand™ Clear Graduated Glass Bottles with Black Phenolic Pulp/Tin Foil Lined Cap


Clear Glass Boston Round Bottles are excellent general use bottles ideal for when content integrity and sample or product visibility is important.

Manufacturer: fisherbrand™  ALP263460

 View more versions of this product

Phenolic Pulp/Tin Foil lined cap

 Alert

Not recommended for acids and alkalis.

Catalog No. 02-911-911A

  • / Case of 24 Each
 

Description & Specifications

Specifications

Type Bottle, Graduated
Closure Color Black
Mouth Wide
Shape Round
Closure Type Screw
Liner Material Pulp/Tin Foil
Neck Finish 70-400
Closure Material Phenolic
Material Glass
Quantity 24/Cs.
Color Clear
Capacity (English) 16 oz.
Capacity (Metric) 480mL

Clear Graduated Glass Bottles

  • Graduated in both milliliters and ounces
  • For use with biological and pathological specimens, but can also be used for storing industrial laboratory chemicals and reagents
  • Glass offers minimal chemical interference with contents; clear (flint) glass is appropriate when visibility is important but protection from UV rays is not necessary
  • Designed with a slight shoulder for easy pouring and handling
  • Wide mouth facilitates easy mixing, handling, and transfer of contents
Pulp/Tin-Foil Lined Phenolic Caps
  • Offer good chemical, impact, and temperature resistance and are typically used with solids and powders
  • Plastic screw caps with pulp/tin-foil liners have strong resistance to hydro-carbons, alcohols, ketones and oils, but are not recommended for acids and alkalis