missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Graduated Glass Bottle Beakers with Black Phenolic PolyCone Cap

Multipurpose containers for use with biological and pathological specimens. Fisherbrand™ Graduated Glass Bottle Beakers with Black Phenolic PolyCone Cap can also be used for storing industrial laboratory chemicals and reagents.

Manufacturer:  Fisherbrand™ ALP263472

 View more versions of this product

Catalog No. 02-911-471A


 

Includes: Phenolic Poly Cone lined cap

Specifications

Description

AC Round, Graduated; Fisherbrand; Clear; With black Phenolic Poly Cone lined cap attached; 2 oz., 60mL; 38-400; 288/Cs.

  • Graduated in both milliliters and ounces
  • Clear (flint) glass bottles are designed with a slight shoulder for easy pouring and handling
  • Wide mouth facilitates easy mixing, handling, and transfer of contents
Description & Specifications

Specifications

60mL
Phenolic
Glass
Bottle, Graduated
PolyCone
38-400
Screw
2 oz.
Clear
288/Cs.
Black
Wide
Round