missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Graduated Glass Bottle Beakers with Black Phenolic PolyCone Cap

Multipurpose containers for use with biological and pathological specimens. Fisherbrand™ Graduated Glass Bottle Beakers with Black Phenolic PolyCone Cap can also be used for storing industrial laboratory chemicals and reagents.

Manufacturer:  Fisherbrand™ ALP263464

 View more versions of this product

Catalog No. 02-911-470A



Includes: Phenolic Poly Cone lined cap

Description

Description

Graduated Medium Rounds are excellent multipurpose containers for use with biological and pathological specimens, but can also be used for storing industrial laboratory chemicals and reagents. These clear (flint) glass bottles are designed with a slight shoulder for easy pouring and handling. The wide mouth facilitates easy mixing, handling, and transfer of contents. Graduated in both milliliters and ounces.

  • Graduated in both milliliters and ounces
  • Clear (flint) glass bottles are designed with a slight shoulder for easy pouring and handling
  • Wide mouth facilitates easy mixing, handling, and transfer of contents

Specifications

Specifications

30mL
Phenolic
Glass
Bottle, Graduated
PolyCone
33-400
Screw
1 oz.
Clear
432/Cs.
Black
Wide
Round
Documents
Promotions

Promotions