Please login to your online account to display your discounted pricing
Fisherbrand™ Graduated Glass Bottle Beakers without Cap
Multipurpose containers for use with biological and pathological specimens. Fisherbrand™ Graduated Glass Bottle Beakers without Cap can also be used for storing industrial laboratory chemicals and reagents.
$115.70 - $298.30
Specifications
Color Clear
Material Glass
Mouth Wide
Shape Round
View More Specs
Products
Catalog Number Mfr. No. Quantity Capacity (English) Capacity (Metric) Neck Finish Price Quantity    
02-911-941A
fisherbrand™
ALP263463
432/Cs. 1 oz. 30mL 33-400 Case of 432 Each for $298.30
02-912-119A
fisherbrand™
ALP263458
48/Cs. 16 oz. 480mL 70-400 Case of 48 Each for $115.70
02-912-256A
fisherbrand™
ALP263479
144/Cs. 4 oz. 120mL 48-400 Case of 144 Each for $156.17
02-912-255A
fisherbrand™
ALP263471
288/Cs. 2 oz. 60mL 38-400 Case of 288 Each for $245.37
02-912-257A
fisherbrand™
ALP263485
96/Cs. 8 oz. 240mL 58-400 Case of 96 Each for $202.73
Description & Specifications
Specifications
Color Clear
Material Glass
Mouth Wide
Shape Round
  • Graduated in both milliliters and ounces
  • Clear (flint) glass bottles are designed with a slight shoulder for easy pouring and handling
  • Wide mouth facilitates easy mixing, handling, and transfer of contents