missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Graduated Glass Bottle Beakers without Cap

Multipurpose containers for use with biological and pathological specimens. Fisherbrand™ Graduated Glass Bottle Beakers without Cap can also be used for storing industrial laboratory chemicals and reagents.

Manufacturer:  Fisherbrand™ ALP263479

 View more versions of this product

Catalog No. 02-912-256A


 

Includes: Clear Graduated AC Round Bottle

Description

Description

Graduated Medium Rounds are excellent multipurpose containers for use with biological and pathological specimens, but can also be used for storing industrial laboratory chemicals and reagents. These clear (flint) glass bottles are designed with a slight shoulder for easy pouring and handling. The wide mouth facilitates easy mixing, handling, and transfer of contents. Graduated in both milliliters and ounces.

  • Graduated in both milliliters and ounces
  • Clear (flint) glass bottles are designed with a slight shoulder for easy pouring and handling
  • Wide mouth facilitates easy mixing, handling, and transfer of contents
Specifications

Specifications

120mL
ClearþClear
144/Cs.
Wide
Round
4 oz.
GlassþGlass
Bottle, GraduatedþGraduated Medium Round Bottle
48-400
Documents