Please login to your online account to display your discounted pricing
Fisherbrand™ Graduated Glass Bottle Beakers without Cap
Multipurpose containers for use with biological and pathological specimens. Fisherbrand™ Graduated Glass Bottle Beakers without Cap can also be used for storing industrial laboratory chemicals and reagents.
Manufacturer: fisherbrand™  ALP263471
 View more versions of this product
Clear Graduated AC Round Bottle
Catalog No. 02-912-255A
  • / Case of 288 Each
Description & Specifications
Specifications
Type Graduated Bottle
Neck Finish 38-400
Shape Round
Mouth Wide
Quantity 288/Cs.
Capacity (Metric) 60mL
Material Glass
Color Clear
Capacity (English) 2 oz.
  • Graduated in both milliliters and ounces
  • Clear (flint) glass bottles are designed with a slight shoulder for easy pouring and handling
  • Wide mouth facilitates easy mixing, handling, and transfer of contents