missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Graduated Glass Bottle Beakers without Cap

Multipurpose containers for use with biological and pathological specimens. Fisherbrand™ Graduated Glass Bottle Beakers without Cap can also be used for storing industrial laboratory chemicals and reagents.

Manufacturer:  Fisherbrand™ ALP263458

 View more versions of this product

Catalog No. 02-912-119A


 

Includes: Clear Graduated AC Round Bottle

Specifications

Description

AC Round, Graduated; Fisherbrand; Clear; Bottle only16 oz., 480mL; 70-400; 48/Cs.

  • Graduated in both milliliters and ounces
  • Clear (flint) glass bottles are designed with a slight shoulder for easy pouring and handling
  • Wide mouth facilitates easy mixing, handling, and transfer of contents
Description & Specifications

Specifications

480mL
Clear
48/Cs.
Wide
Round
16 oz.
Glass
Bottle, Graduated
70-400