Please login to your online account to display your discounted pricing
Fisherbrand™ Clear French Square Bottles with Black Phenolic PolyCone Cap
Ideal for mixing, storing and sampling. Fisherbrand™ Clear French Square Bottles with Black Phenolic PolyCone Cap are spcace-saving and easy to fill.
$63.60 - $403.00
Specifications
Closure Color Black
Closure screw cap
Closure Material Phenolic
Closure Liner Poly-seal® (polycone)
Closure Type Screw
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Neck Finish Price Quantity    
02-911-403
fisherbrand™
263964
30mL 24-400 Case of 48 Each for $145.00
02-911-402
fisherbrand™
263956
15mL 20-400 Case of 48 Each for $107.47
02-911-404
fisherbrand™
263967
60mL 28-400 Case of 48 Each for $106.47
02-911-405
fisherbrand™
263973
120mL 33-400 Case of 24 Each for $63.60
02-911-875
fisherbrand™
W014125
120mL 33-400 Case of 120 Each for $403.00
Description & Specifications
Specifications
Closure Color Black
Closure screw cap
Closure Material Phenolic
Closure Liner Poly-seal® (polycone)
Closure Type Screw
Color Clear
Material Glass
Liner Material PolyCone
Mouth Wide
Shape French Square
Closure Included Yes
  • Clear glass provides maximum content visibility and minimal chemical inference with bottle's contents
  • French Square bottle design allows bottle to hold maximum volume of contents while taking up minimum shelf and storage space