Fisherbrand™ Cleanroom Pens
Designed with low sodium ink to prevent contamination in cleanroom settings.  Fisherbrand™ Cleanroom Pens come in black, blue, and red and feature HDPE construction.
Supplier:  Fisherbrand™ 18001508
 View more versions of this product
Catalog No. 18-001-508
Description
Description
  • Manufactured with very low sodium inks to prevent contamination
  • Barrel and cap made from high density polyethylene (HDPE)
  • Pens are packaged 10 per pack
  • Available in Black (18-001-507), Blue (18-001-511), or Red (18-001-508)
Specifications
Specifications
Red