Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Clathrin, Light Chain Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments.

$244.80 - $1,219.29

Specifications

Antigen Clathrin, Light Chain Ab-1
Clone CON.1
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS1291P

 
thermo scientific™ lab vision™
MS1291P
Purified Ab with BSA and Azide 1mL Each for $1,121.98

MS1291P0

 
thermo scientific™ lab vision™
MS1291P0
Purified Ab with BSA and Azide 100µL Each for $244.80

MS1291P1

 
thermo scientific™ lab vision™
MS1291P1
Purified Ab with BSA and Azide 500µL Each for $705.00

MS1291P1ABX

 
thermo scientific™ lab vision™
MS1291P1ABX
Ab without BSA/Azide (Frozen) 100µL Each for $783.71

MS1291PABX

 
thermo scientific™ lab vision™
MS1291PABX
Ab without BSA/Azide (Frozen) 200µL Each for $1,219.29

MS1291R7

 
thermo scientific™ lab vision™
MS1291R7
Ready-to-use for IHC (UVLP) 7mL Each for $769.04
Description & Specifications

Specifications

Antigen Clathrin, Light Chain Ab-1
Clone CON.1
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Bovine
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline Cytoskeletal Protein
Type Primary
Immunogen Synthetic peptide derived from bovine clathrin light chain corresponding to the conserved region of the light chains, residues 23-44
Isotype IgG2b kappa

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ Clathrin, Light Chain Ab-1, Mouse Monoclonal Antibody.

Clathrin consists of three heavy chain (192 kDa) and three light chains (LCa or LCb, 25-29 kDa) and self-assembles into a polyhedral cytoskeletal element. Self-assembly facilitates clathrin's central role in membrane sorting and selective transport within the cell.

Host Species: Mouse

Clone: CON.1

Isotype: IgG2b/κ

Species Reactivity: Human and Cow. Others not known.

Epitope: aa 23-44

Immunogen: Synthetic peptide derived from bovine clathrin light chain corresponding to the conserved region of the light chains, residues 23-44

Molecular Weight: 25-29 kDa

Positive Control: Placenta

Cellular Localization: Membrane and cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO