missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Clathrin, Light Chain Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS1291PABX

 View more versions of this product

Catalog No. MS1291PABX


 

Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ Clathrin, Light Chain Ab-1, Mouse Monoclonal Antibody.

Clathrin consists of three heavy chain (192 kDa) and three light chains (LCa or LCb, 25-29 kDa) and self-assembles into a polyhedral cytoskeletal element. Self-assembly facilitates clathrin's central role in membrane sorting and selective transport within the cell.

Host Species: Mouse

Clone: CON.1

Isotype: IgG2b/κ

Species Reactivity: Human and Cow. Others not known.

Epitope: aa 23-44

Immunogen: Synthetic peptide derived from bovine clathrin light chain corresponding to the conserved region of the light chains, residues 23-44

Molecular Weight: 25-29 kDa

Positive Control: Placenta

Cellular Localization: Membrane and cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Clathrin, Light Chain Ab-1
Monoclonal
1mg/mL
Ab without BSA/Azide (Frozen)
Murine
IgG2b kappa
RUO
Primary
Bovine,Human
Immunofluorescence,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blotting
CON.1
Unlabeled
Concentrated
Synthetic peptide derived from bovine clathrin light chain corresponding to the conserved region of the light chains, residues 23-44
200μL
Cytoskeletal Protein
Monoclonal