Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Clathrin, Light Chain Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments.

Manufacturer: thermo scientific™ lab vision™  MS1291P1ABX

 View more versions of this product

Catalog No. MS1291P1ABX

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Synthetic peptide derived from bovine clathrin light chain corresponding to the conserved region of the light chains, residues 23-44
Cross Reactivity Bovine
Cross Reactivity Human
Antigen Clathrin, Light Chain Ab-1
Regulatory Status RUO
Clone CON.1
Quantity 100µL
Isotype IgG2b kappa
Format Concentrated
Classification Monoclonal
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Cytoskeletal Protein
Concentration 1mg/mL
Conjugate Unlabeled
Type Primary

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments with Thermo Scientific™ Clathrin, Light Chain Ab-1, Mouse Monoclonal Antibody.

Clathrin consists of three heavy chain (192 kDa) and three light chains (LCa or LCb, 25-29 kDa) and self-assembles into a polyhedral cytoskeletal element. Self-assembly facilitates clathrin's central role in membrane sorting and selective transport within the cell.

Host Species: Mouse

Clone: CON.1

Isotype: IgG2b/κ

Species Reactivity: Human and Cow. Others not known.

Epitope: aa 23-44

Immunogen: Synthetic peptide derived from bovine clathrin light chain corresponding to the conserved region of the light chains, residues 23-44

Molecular Weight: 25-29 kDa

Positive Control: Placenta

Cellular Localization: Membrane and cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO