missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Classifying Life Poster

See the similarities and differences of life on Earth.

Manufacturer:  Fisher Scientific AZ351066

 View more versions of this product

Catalog No. S67938



Description

Description

  • Stunning photographs and detailed illustrations provide your students with a stimulating view
  • Dimensions: 23 × 35 in. (57.5 × 87.5cm)

Specifications

Specifications

Life science
35 in.
Representative organisms from the archaebacteria, eubacteria, protista, fungi, plantae and animal kingdoms
Classifying Life Poster
58.42cm
Provide students with a stimulating view of the similarities and differences of life on earth
88.9cm
Laminated
23 in.
Documents