Please login to your online account to display your discounted pricing

Classifying Life Poster


See the similarities and differences of life on Earth.

Manufacturer: fisher science education  AZ-35-1065

 View more versions of this product

Catalog No. S67937

  • / Each
 

Description & Specifications

Specifications

Product Type Classifying Life Poster
For Use With (Application) Provide students with a stimulating view of the similarities and differences of life on earth
Height (English) 35 in.
Height (Metric) 88.9cm
Includes Representative organisms from the archaebacteria, eubacteria, protista, fungi, plantae and animal kingdoms
Width (English) 23 in.
Width (Metric) 58.42cm
Classification Life science
Material Paper