Please login to your online account to display your discounted pricing

Classification of Soil Layers

Discover the conditions that caused your local soils to form.

$59.40 - $59.40

Specifications

Certifications/Compliance NSES: A, D, F, G
For Use With (Application) Determine how soils are formed and create a sample soil profile
Grade 7 to 12
Sufficient For 40 students
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    

S01407

 
fisher science education
20-4143
Classification of soil layers lab investigation Each for $59.40
Description & Specifications

Specifications

Certifications/Compliance NSES: A, D, F, G
For Use With (Application) Determine how soils are formed and create a sample soil profile
Grade 7 to 12
Sufficient For 40 students

  • Determine how soils are formed
  • Create a sample soil profile using the provided soil horizon cards
  • Determine the environmental changes that cause those soils to form
  • Portable and practical soil cards

Grades 7 to 12. Class size: 40 students.

NSES: A, D, F, G