Binder™ Waterproof Interior Socket
Binder™ Waterproof Interior Socket for use with a range of Binder™ incubators, growth chambers, freezers, drying and heating chambers and climate chambers.
$1,605.73 - $1,730.54
Specifications
Description CA3GD mounting box
Includes Cover and corresponding plug
Type Interior Socket
Wattage Max. 500 w
View More Specs
Products 6
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
08111243 View Documents Binder™
80121005
KT 115 Each for $1,605.73
08111244 View Documents Binder™
80121006
KT 170 Each for $1,605.73
08111245 View Documents Binder™
80121171
KB 23 Each for $1,648.15
08111246 View Documents Binder™
80121074
KB 53, KB 115 Each for $1,648.15
08111247 View Documents Binder™
80121075
KB 720, KB 240, KB 400 Each for $1,648.15
08111248 View Documents Binder™
80121043
KBF 240, KBF-S 240, KBF 720, KBF-S 720, KBF 1020, KBF 115 Each for $1,730.54
Description
Description
  • With cover and corresponding plug
  • Switched flush-mounting box (CA3GD) for nominal voltage in unit interior
  • Protection class of IP67
  • Maximum temperature of 90°C
Specifications
Specifications
CA3GD mounting box
Cover and corresponding plug
Interior Socket
Max. 500 w