missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Class A Glass Burettes with PTFE Rota Flow Stopcock

Eisco™ Class A Glass Burettes with PTFE Rota Flow Stopcock designed with blue graduations and a straight single bore.

$32.68 - $51.03

Specifications

Material Borosilicate Glass
For Use With (Application) Dispensing Liquid
Scale Blue
Stopcock Straight
Stopcock Material PTFE
View More Specs

Products 4
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
S29400
View Documents
Eisco™
CH0238A
10 mL
Each for $32.68
 
S29401
View Documents
Eisco™
CH0238B
25 mL
Each for $35.94
 
S29402
View Documents
Eisco™
CH0238C
50 mL
Each for $43.71
 
S29403
View Documents
Eisco™
CH0238D
100 mL
Each for $51.03
 
Description

Description

  • Made from high quality 3.3 borosilicate glass
  • Blue ceramic graduations
  • Straight single bore
  • Includes PTFE Rota flow stopcock
  • Needle valve stopcock

Warranty

1 year limited manufacturer warranty

Specifications

Specifications

Borosilicate Glass
Blue
PTFE
Class A
Dispensing Liquid
Straight
Straight Bore Buret
SDS
missing translation for 'documents'

missing translation for 'documents'