missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Binder™ Class 3.3 Independent Temperature Safety Device

Binder™ Class 3.3 Independent Temperature Safety Device for safe and consistent temperature regulation.

$2,264.98 - $3,078.56

Specifications

Description Class 3.3
For Use With (Application) Temperature regulation
Includes Visual alarm
Type Temperature Safety Device
View More Specs

Products 15
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
08111173 View Documents Binder™
80120999
KT 115 Each for $2,264.98
 
08111174 View Documents Binder™
80121000
KT 170 Each for $2,264.98
 
08111172 View Documents Binder™
80120978
KT 53 Each for $2,264.98
 
08111175 View Documents Binder™
80121080
KB 53, KB 115 Each for $2,334.38
 
08111176 View Documents Binder™
80121081
KB 720, KB 240, KB 400 Each for $2,931.96
 
08111177 View Documents Binder™
80121032
KBF 115 Each for $3,078.56
 
08111178 View Documents Binder™
80121033
KBF 240 Each for $3,078.56
 
08111179 View Documents Binder™
80121034
KBF 1020, KBF 720 Each for $3,078.56
 
08111180 View Documents Binder™
80121751
KBF P 240 Each for $3,078.56
 
08111181 View Documents Binder™
80121754
KBF P 720 Each for $3,078.56
 
08111182 View Documents Binder™
80121752
KBF LQC 240 Each for $3,078.56
 
08111183 View Documents Binder™
80121755
KBF LQC 720 Each for $3,078.56
 
08111184 View Documents Binder™
80121673
KMF 115 Each for $3,078.56
 
08111185 View Documents Binder™
80121674
KMF 240 Each for $3,078.56
 
08111186 View Documents Binder™
80121675
KMF 720 Each for $3,078.56
 
Description

Description

  • Class 3.3
  • With visual alarm
  • Safety devices also available in Class 2 and 3.1
Specifications

Specifications

Class 3.3
Temperature regulation
Visual alarm
Temperature Safety Device