missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BINDER™ Class 3.3 Independent Temperature Safety Device
For Use With: KMF 240
Supplier: BINDER™ 80121674
Description
- Class 3.3
- With visual alarm
- Safety devices also available in Class 2 and 3.1
Specifications
Class 3.3 independent temperature safety device - with visual alarm (DIN 12880) | |
Temperature Safety Device | |
Visual alarm |
BINDER Chambers KMF 240 | |
Temperature Regulation |