missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Clarithromycin, 960-1040 ^mg/mg, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J66005MC
Description
A macrolide antibiotic that inhibits bacterial growth by targeting the 50S ribosomal subunit
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- A macrolide antibiotic that inhibits bacterial growth by targeting the 50S ribosomal subunit
Specifications
Clarithromycin | |
960 to 1040μg/mg | |
81103-11-9 | |
C38H69NO13 | |
14,2339 | |
AGOYDEPGAOXOCK-KCBOHYOISA-N | |
(3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-14-ethyl-12,13-dihydroxy-4-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-7-methoxy-3,5,7,9,11,13-hexamethyl-oxacyclotetradecane-2,10-dion | |
84029 | |
747.95 | |
100 mg | |
86°C |
Clarithromycin | |
Powder | |
217°C to 220°C | |
MFCD00865140 | |
Soluble in DMSO | |
CCC1C(C(C(C(=O)C(CC(C(C(C(C(C(=O)O1)C)OC2CC(C(C(O2)C)O)(C)OC)C)OC3C(C(CC(O3)C)N(C)C)O)(C)OC)C)C)O)(C)O | |
747.964 | |
CHEBI:3732 | |
White | |
>110°C (230°F) |
Chemical Identifiers
81103-11-9 | |
747.964 | |
AGOYDEPGAOXOCK-KCBOHYOISA-N | |
CHEBI:3732 | |
CCC1C(C(C(C(=O)C(CC(C(C(C(C(C(=O)O1)C)OC2CC(C(C(O2)C)O)(C)OC)C)OC3C(C(CC(O3)C)N(C)C)O)(C)OC)C)C)O)(C)O |
C38H69NO13 | |
MFCD00865140 | |
84029 | |
(3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-14-ethyl-12,13-dihydroxy-4-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-7-methoxy-3,5,7,9,11,13-hexamethyl-oxacyclotetradecane-2,10-dion |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
RTECSNumber : KF4997000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only