Please login to your online account to display your discounted pricing

Sartorius Claristep™ Filter, 0.2μm

Make light work of a tedious task. Use Claristep™ filter units to remove particles from samples prior to analytics. The filters are suitable for small sample volumes of 60 to 600μL and are used without syringe. Syringeless filters and simple manual processing simplify sample clarification.

$300.00 - $1,278.00

Specifications

For Use With (Application) HPLC (high-performance liquid chromatography)
Pore Size 0.2μm
Storage Requirements Room Ambient
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

14-556-010

 
sartorius
17C07FT96
96 pk Pack for $300.00

14-556-011

 
sartorius
17C07FT480
480 pk Pack for $1,278.00
Description & Specifications

Specifications

For Use With (Application) HPLC (high-performance liquid chromatography)
Pore Size 0.2μm
Storage Requirements Room Ambient

    Features
  • Process samples without a vacuum source or a power supply
  • Multiplexing of up to 8 samples
  • Syringeless
  • Suitable for low sample volumes
  • Non-sterile
  • Hydrophilic
  • Diameter: 9.7mm
  • Membrane Filter Material: Regenerated cellulose (RC)
  • Housing Material: Polypropylene (PP) Housing