Please login to your online account to display your discounted pricing

Thermo Scientific™ CL-XPosure™ Film (X-Ray Film)

Capture results of ECL-based Western blots and other chemiluminescent protein or nucleic acid assays using this economically priced, clear-blue X-ray film.

Manufacturer: thermo scientific™  34088

 View all options for this product

Catalog No. PI34088


Description & Specifications


Description CL-XPosure Film, 5 x 7in. (12.5 x 17.5cm)
Formulation Blue-backed, silver-emulsion, two-sided autoradiography film
Quantity 50 sheets
Sufficient For 50 uses

Thermo Scientific™ CL-XPosure Film is an economically priced clear-blue X-ray film especially for detection and documentation of ECL-based Western blots and other chemiluminescent protein or nucleic acid assay methods.

CL-XPosure Film is an excellent phototgraphic film for use with enhanced chemiluminescence (ECL) substrates for horseradish peroxidase (HRP) or alkaline phosphatase (AP). Convenient sizes are offered for protein Western blotting, standard film cassettes and film developing equipment. The autoradiography film also works well for chemiluminescent or isotopic DNA and RNA methods, including Southern and Northern blotting and gel-shift assays (EMSA). The quality and price of this film make it the smart choice for life science research applications.


  • High performance – just as sensitive as other commercially available autoradiography films

  • Affordable – costs up to 75% less than other brands of comparable quality X-ray film

  • Convenient – available in five sheet sizes in packages containing 50 or 100 non-interleaved sheets

  • Perfect match – validated for use with Thermo Scientific SuperSignal Chemiluminescent Substrates and Thermo Scientific Pierce ECL Chemiluminescent Substrate

Ordering Information:

Available in 5 × 7in., 7 × 9.5in. , 9.5 × 11in., or 14 × 17in. sheets