Please login to your online account to display your discounted pricing

CL 316243 disodium salt, Tocris

Highly selective β3 agonist

$233.76 - $963.39

Chemical Identifiers

Chemical Name or Material CL 316243 disodium salt
Assay Percent Range >96%
Molecular Formula C20H18ClNNa2O7
Formula Weight 465.8
Synonym 5-[(2R)-2-[[(2R)-2-(3-Chlorophenyl)-2-hydroxyethyl]amino]propyl]-1, 3-benzodioxole-2, 2-dicarboxylic acid disodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

149950

 
r & d systems
RDS149950
50mg Each for $963.39

14-991-0

 
r & d systems
RDS149910
10mg Each for $233.76
Description & Specifications

Specifications

Formula Weight 465.8
Quantity 50mg
Chemical Name or Material CL 316243 disodium salt
Molecular Formula C20H18ClNNa2O7
CAS 151126-84-0
Synonym 5-[(2R)-2-[[(2R)-2-(3-Chlorophenyl)-2-hydroxyethyl]amino]propyl]-1, 3-benzodioxole-2, 2-dicarboxylic acid disodium salt
Assay Percent Range >96%
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C