Please login to your online account to display your discounted pricing

CL 218872, Tocris

Benzodiazepine agonist

$121.23 - $441.50

Chemical Identifiers

Chemical Name or Material CL 218872
Assay Percent Range >99%
Molecular Formula C13H9F3N4
Formula Weight 278.24
Synonym 3-Methyl-6-[-3-(trifluoromethyl)phenyl]-1,2,4-triazolo[4,3-b]pyridazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

170910

 
r & d systems
170910
10mg Each for $121.23

170950

 
r & d systems
170950
50mg Each for $441.50
Description & Specifications

Specifications

Formula Weight 278.24
Quantity 10mg
Assay Percent Range >99%
CAS 66548-69-4
Molecular Formula C13H9F3N4
Chemical Name or Material CL 218872
Synonym 3-Methyl-6-[-3-(trifluoromethyl)phenyl]-1,2,4-triazolo[4,3-b]pyridazine
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature