Please login to your online account to display your discounted pricing

Tocris™ CK 869

Arp2/3 inhibitor; inhibits actin polymerization

$225.77 - $924.23

Specifications

Product Type CK 869
Inhibitors CK 869
Molecular Weight 394.28
Purity >98
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

498410R

 
tocris™
RDS498410
10mg Each for $225.77

498450R

 
tocris™
RDS498450
50mg Each for $924.23
Description & Specifications

Specifications

Product Type CK 869
Inhibitors CK 869
Molecular Weight 394.28
Purity >98

Inhibitor of human and bovine actin-related protein 2/3 (Arp2/3) complex. Inhibits actin polymerization (IC50= 11μM). Does not inhibit budding or fission yeast Arp2/3 complexes.

  • Chemical Name: 2-(3-Bromophenyl)-3-(2,4-dimethoxyphenyl)-4-thiazolidinone
  • Formula: C17H16BrNO3S
  • CAS Number: 388592-44-7
  • Solubility: Soluble to 100mM in DMSO
  • Purity: >98