Please login to your online account to display your discounted pricing

CJ 033466, Tocris

Selective 5-HT4 partial agonist

$188.60 - $794.96

Chemical Identifiers

Chemical Name or Material CJ 033466
Assay Percent Range >98%
Molecular Formula C19H28ClN5O
Formula Weight 377.91
Synonym 5-Amino-6-chloro-2-methyl-N-[[1-(2-methylpropyl)-4-piperidinyl]methyl]-imidazo[1, 2-a]pyridine-8-carboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

308950

 
r & d systems
RDS308950
50mg Each for $794.96

308910

 
r & d systems
RDS308910
10mg Each for $188.60
Description & Specifications

Specifications

Formula Weight 377.91
Quantity 50mg
Synonym 5-Amino-6-chloro-2-methyl-N-[[1-(2-methylpropyl)-4-piperidinyl]methyl]-imidazo[1, 2-a]pyridine-8-carboxamide
CAS 519148-48-2
Molecular Formula C19H28ClN5O
Chemical Name or Material CJ 033466
Assay Percent Range >98%
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C