missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Citronellol 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 110465000

 View more versions of this product

Catalog No. AC110465000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥94%
Liquid
Clear almost colorless
01, 451
1.4550 to 1.4580 (20°C, 589nm)
β-Citronellol
Citronellol
(CH3)2C=CHCH2CH2CH(CH3)CH2CH2OH
95%
Authentic
156.27
500mL
15, 2329
0.852 to 0.856 (20°C)
10mmol/L max.
Glass bottle
95%
C10H20O
MFCD00002935
106-22-9

Chemical Identifiers

106-22-9
C10H20O
156.27
β-Citronellol
≥94%
95%
(CH3)2C=CHCH2CH2CH(CH3)CH2CH2OH
MFCD00002935
Citronellol, 95%

Safety and Handling

SDS & Safety Documents

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 203-375-0

RTECSNumber : RH3400000

TSCA : TSCA