Learn More
Citronellic acid, 94%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2286222
Description
Citronellic acid was used in synthesis of stereoisomers of 3,11- dimethylnonacosan-2-one and 29-hydroxy-3,11-dimethylnonacosan-2-one.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCitronellic acid was used in synthesis of stereoisomers of 3,11- dimethylnonacosan-2-one and 29-hydroxy-3,11-dimethylnonacosan-2-one.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Citronellic acid | |
0.923 | |
>110°C (230°F) | |
1.4535 | |
100 g | |
citronellic acid, 3,7-dimethyl-6-octenoic acid, rhodinolic acid, citronellate, rhodinic acid, 6-octenoic acid, 3,7-dimethyl, fema no. 3142, citronellicacid, callitrol, acmc-20apjr | |
GJWSUKYXUMVMGX-UHFFFAOYNA-N | |
3,7-dimethyloct-6-enoic acid | |
10402 | |
170.25 |
502-47-6 | |
121°C to 122°C (1 mmHg) | |
C10H18O2 | |
MFCD00002728 | |
UN2810 | |
Insoluble in water. | |
CC(CCC=C(C)C)CC(O)=O | |
170.25 | |
CHEBI:80507 | |
94% |
Chemical Identifiers
502-47-6 | |
170.25 | |
GJWSUKYXUMVMGX-UHFFFAOYNA-N | |
10402 | |
3,7-dimethyloct-6-enoic acid |
C10H18O2 | |
MFCD00002728 | |
citronellic acid, 3,7-dimethyl-6-octenoic acid, rhodinolic acid, citronellate, rhodinic acid, 6-octenoic acid, 3,7-dimethyl, fema no. 3142, citronellicacid, callitrol, acmc-20apjr | |
CHEBI:80507 | |
CC(CCC=C(C)C)CC(O)=O |
Safety and Handling
GHS H Statement
H312-H315-H319-H335
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P280-P302+P352-P312-P363-P501c
H312
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 207-939-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only