Learn More
Citronellal, 93%, pract., Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 405291000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Citronellal | |
106-23-0 | |
0.8600g/mL | |
78°C | |
91% min. (GC) | |
C10H18O | |
(CH3)2CCHCH2CH2CH(CH3)CH2CHO | |
100 g | |
15, 2328 | |
Solubility in water: insoluble. Other solubilities: soluble in organic solvents,ethanol,ether | |
C[C@H](CCC=C(C)C)CC=O | |
154.25 | |
CHEBI:47856 | |
93% |
pract., 93% | |
Yellow | |
206°C to 208°C | |
Authentic | |
Glass bottle | |
1.4440 to 1.45 | |
MFCD00038090 | |
0.86 | |
citronellal, rhodinal, 3,7-dimethyl-6-octenal, 2,3-dihydrocitral, 6-octenal, 3,7-dimethyl, citronellel, citronella, 3,7-dimethyl-6-octen-1-al, beta-citronellal, d-rhodinal | |
NEHNMFOYXAPHSD-SNVBAGLBSA-N | |
3,7-dimethyloct-6-enal | |
7794 | |
154.25 | |
Liquid |
Chemical Identifiers
106-23-0 | |
154.25 | |
NEHNMFOYXAPHSD-SNVBAGLBSA-N | |
7794 | |
3,7-dimethyloct-6-enal |
C10H18O | |
MFCD00038090 | |
citronellal, rhodinal, 3,7-dimethyl-6-octenal, 2,3-dihydrocitral, 6-octenal, 3,7-dimethyl, citronellel, citronella, 3,7-dimethyl-6-octen-1-al, beta-citronellal, d-rhodinal | |
CHEBI:47856 | |
C[C@H](CCC=C(C)C)CC=O |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause an allergic skin reaction.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation or rash occurs: G
GHS Signal Word: Warning
EINECSNumber : 203-376-6
RTECSNumber : RH2140000
TSCA : TSCA
RUO – Research Use Only