Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-Citronellal, 96%

$51.23 - $126.88

Chemical Identifiers

Chemical Name or Material (+/-)-Citronellal
CAS 106-23-0
Molecular Formula C10H18O
Formula Weight 154.25
MDL Number MFCD00038090
Beilstein 1720789
Merck Index 14,2329
Synonym (+/-)-3,7-Dimethyl-6-octenal
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL15753AE

  SDS
alfa aesar™
L15753AE
100mL Each for $51.23

AAL15753AP

  SDS
alfa aesar™
L15753AP
500mL Each for $126.88
Description & Specifications

Specifications

Formula Weight 154.25
Beilstein 1720789
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number RH2140000
Synonym (+/-)-3,7-Dimethyl-6-octenal
Merck Index 14,2329
CAS 106-23-0
Flash Point 75°C (167°F)
Quantity 100mL
Assay 96%
Health Hazard 2 GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Molecular Formula C10H18O
Chemical Name or Material (+/-)-Citronellal
Boiling Point 206° to 208°C
TSCA TSCA
MDL Number MFCD00038090

Applications
(^+)-Citronellal is an active component of citronella oil. It is used as a flavoring agent, insect repellent, fragrance in soaps and detergents, and in personal care products like creams, lotions, and perfumes. It acts as a chemical intermediate for citronellol and hydroxycitronellal. Further, it is used to prepare 1-dibutylamino-3,7-dimethyl-oct-6-en-2-one by reacting with dibutylamine. In addition to this, it is utilized to inhibit the development of mechanical nociception induced by carrageenan (CG) and tumor necrosis factor-alfa.

Solubility
Slightly miscible with water and ethanol.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Air and light sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.

Safety and Handling

Warning

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.