missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Citrinin, 98%, ACROS Organics™

$275.54 - $275.54

Chemical Identifiers

CAS 518-75-2
Molecular Formula C13H14O5
Molecular Weight (g/mol) 250.25
MDL Number MFCD00006912
InChI Key CBGDIJWINPWWJW-IYSWYEEDSA-N
Synonym citrinin, antimycin, --citrinin, citriain, 3r,4s-4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-3h-2-benzopyran-7-carboxylic acid, unii-3s697x6snz, ccris 175, 3h-2-benzopyran-7-carboxylic acid, 4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-, 3r-trans, 3r-trans-4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-3h-2-benzopyran-7-carboxylic acid, 4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-3h-2-benzopyran-7-carboxylic acid
PubChem CID 54680783
IUPAC Name (3R,4S)-6-hydroxy-3,4,5-trimethyl-8-oxo-3,4-dihydroisochromene-7-carboxylic acid
SMILES CC1C(OC=C2C1=C(C(=C(C2=O)C(=O)O)O)C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC229580050 SDS View Documents Acros Organics
229580050
5mg Each for $275.54
Specifications

Chemical Identifiers

518-75-2
250.25
CBGDIJWINPWWJW-IYSWYEEDSA-N
54680783
CC1C(OC=C2C1=C(C(=C(C2=O)C(=O)O)O)C)C
C13H14O5
MFCD00006912
citrinin, antimycin, --citrinin, citriain, 3r,4s-4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-3h-2-benzopyran-7-carboxylic acid, unii-3s697x6snz, ccris 175, 3h-2-benzopyran-7-carboxylic acid, 4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-, 3r-trans, 3r-trans-4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-3h-2-benzopyran-7-carboxylic acid, 4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-3h-2-benzopyran-7-carboxylic acid
(3R,4S)-6-hydroxy-3,4,5-trimethyl-8-oxo-3,4-dihydroisochromene-7-carboxylic acid

Specifications

Authentic
− 30.30
5mg
98%
MFCD00006912
citrinin, antimycin, --citrinin, citriain, 3r,4s-4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-3h-2-benzopyran-7-carboxylic acid, unii-3s697x6snz, ccris 175, 3h-2-benzopyran-7-carboxylic acid, 4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-, 3r-trans, 3r-trans-4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-3h-2-benzopyran-7-carboxylic acid, 4,6-dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-3h-2-benzopyran-7-carboxylic acid
CC1C(OC=C2C1=C(C(=C(C2=O)C(=O)O)O)C)C
250.25
250.25
98%
Glass bottle
Yellow
518-75-2
C13H14O5
15, 2326
CBGDIJWINPWWJW-IYSWYEEDSA-N
(3R,4S)-6-hydroxy-3,4,5-trimethyl-8-oxo-3,4-dihydroisochromene-7-carboxylic acid
54680783
Crystalline Powder or Flakes
Citrinin, 98%
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Toxic in contact with skin.
Causes serious eye irritation.
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 208-257-2

RTECSNumber : DJ2275000