Please login to your online account to display your discounted pricing

Alfa Aesar™ Citrinin, 98%

Induces mitochondrial permeability

Chemical Identifiers

Chemical Name or Material Citrinin
CAS 518-75-2
Molecular Formula C13H14O5
Formula Weight 250.25g/mol
MDL Number MFCD00006912
UN Number UN3462
Beilstein 5282243
Merck Index 14,2327
Synonym 8-Hydroxy-3,4,5-trimethyl-6-oxo-4,6-dihydro-3H-isochromene-7-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ64007LB0

 
alfa aesar™
J64007-LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Yellow
Quantity 5mg
Chemical Name or Material Citrinin
CAS 518-75-2
Physical Form Powder
Assay 98%
Molecular Formula C13H14O5
Formula Weight 250.25g/mol
MDL Number MFCD00006912
UN Number UN3462
Beilstein 5282243
Merck Index 14,2327
Synonym 8-Hydroxy-3,4,5-trimethyl-6-oxo-4,6-dihydro-3H-isochromene-7-carboxylic acid
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331-H351
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing cancer.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXINS, EXTRACTED FROM LIVING SOURCES, SOLID, N.O.S.
RTECS Number DJ2275000
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331-H351
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing cancer.

2° to 8°C